missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leucomalachite Green, pure

CAS: 129-73-7 | C23H26N2 | 330.475 g/mol

$82.61 - $82.61

Chemical Identifiers

CAS 129-73-7
Molecular Formula C23H26N2
Molecular Weight (g/mol) 330.475
MDL Number MFCD00008315
InChI Key WZKXBGJNNCGHIC-UHFFFAOYSA-N
Synonym 4, 4';-Benzylidenebis(N, N-dimethylaniline), Malachite green leuco base
PubChem CID 67215
IUPAC Name 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC402110050
SDS View Documents
Thermo Scientific Chemicals
402110050
5 g Glass bottle
Each for $82.61
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

129-73-7
330.475
WZKXBGJNNCGHIC-UHFFFAOYSA-N
67215
CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C
C23H26N2
MFCD00008315
4, 4';-Benzylidenebis(N, N-dimethylaniline), Malachite green leuco base
4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline

Specifications

129-73-7
MFCD00008315
15, 5763
WZKXBGJNNCGHIC-UHFFFAOYSA-N
CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C
330.475
330.46
Glass bottle
Blue or Brown/Green to White
Powder
C23H26N2
13, 275
4, 4';-Benzylidenebis(N, N-dimethylaniline), Malachite green leuco base
Authentic
4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline
67215
Pure
Complies
5 g
Leucomalachite Green
Videos
Safety and Handling

Safety and Handling

GHS P Statement Suspected of causing cancer.

GHS P Statement Use personal protective equipment as required.

Warning

EINECSNumber : 204-961-9

RTECSNumber : CY1203055

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only