missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leucomalachite Green, pure

Catalog No. AC402110050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402110050 5 g Glass bottle
AC402110250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC402110050 Supplier Thermo Scientific Chemicals Supplier No. 402110050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 129-73-7 | C23H26N2 | 330.475 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 129-73-7
Molecular Formula C23H26N2
Molecular Weight (g/mol) 330.475
MDL Number MFCD00008315
InChI Key WZKXBGJNNCGHIC-UHFFFAOYSA-N
Synonym 4, 4';-Benzylidenebis(N, N-dimethylaniline), Malachite green leuco base
PubChem CID 67215
IUPAC Name 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C

Specifications

Beilstein 13, 275
Merck Index 15, 5763
Infrared Spectrum Authentic
Formula Weight 330.46
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Blue or Brown/Green to White
Quantity 5 g
Physical Form Powder
Chemical Name or Material Leucomalachite Green
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement Suspected of causing cancer.
Precautionary Statement GHS P Statement Use personal protective equipment as required.
EINECSNumber 204-961-9
RTECSNumber CY1203055
TSCA TSCA

RUO – Research Use Only