missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leucomalachite Green

Catalog No. AA4375809
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4375809 10 g
AA4375804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4375809 Supplier Thermo Scientific Chemicals Supplier No. 04375809

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 129-73-7 | C23H26N2 | 330.475 g/mol

Leucomalachite Green is a reagent for the quantitative colorimetric micro determination of hemoglobin and other heme compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 129-73-7
Molecular Formula C23H26N2
Molecular Weight (g/mol) 330.475
MDL Number MFCD00008315
InChI Key WZKXBGJNNCGHIC-UHFFFAOYSA-N
Synonym 4,4'-Benzylidenebis(N,N-dimethylaniline); Malachite green leuco base
PubChem CID 67215
IUPAC Name 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C

Specifications

Melting Point 100°C to 102°C
Sensitivity Air Sensitive
Beilstein 2140506
Merck Index 14,5699
Solubility Information Soluble in water (<0.1mg/ml),chloroform,benzene,ethyl ether,ethanol and toluene.
Quantity 10 g
Physical Form Powder
Formula Weight 330.47
Chemical Name or Material Leucomalachite Green
Hazard Category H341-H351
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 204-961-9
RTECSNumber CY1203055
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only