missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Leucomalachite Green
Description
Leucomalachite Green is a reagent for the quantitative colorimetric micro determination of hemoglobin and other heme compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C to 102°C |
| Sensitivity | Air Sensitive |
| Beilstein | 2140506 |
| Merck Index | 14,5699 |
| Solubility Information | Soluble in water (<0.1mg/ml),chloroform,benzene,ethyl ether,ethanol and toluene. |
| Quantity | 2 g |
| Physical Form | Powder |
| Formula Weight | 330.47 |
| Chemical Name or Material | Leucomalachite Green |
Safety and Handling
| Hazard Category | H341-H351 |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 204-961-9 |
| RTECSNumber | CY1203055 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only