missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leuco Malachite Green 98.0+%, TCI America™
$55.92 - $633.78
Chemical Identifiers
| CAS | 129-73-7 |
|---|---|
| Molecular Formula | C23H26N2 |
| Molecular Weight (g/mol) | 330.475 |
| MDL Number | MFCD00008315 |
| InChI Key | WZKXBGJNNCGHIC-UHFFFAOYSA-N |
| PubChem CID | 67215 |
| IUPAC Name | 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline |
| SMILES | CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C |
Chemical Identifiers
| 129-73-7 | |
| 330.475 | |
| WZKXBGJNNCGHIC-UHFFFAOYSA-N | |
| 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline |
| C23H26N2 | |
| MFCD00008315 | |
| 67215 | |
| CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C |
Specifications
| 102°C | |
| C23H26N2 | |
| 4,4′C-Benzylidenebis(N,N-dimethylaniline), 4,4′C-Bis(dimethylamino)triphenylmethane, 4,4′C-Tetramethyldiaminotriphenylmethane | |
| CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C | |
| 330.475 | |
| 330.48 | |
| 1 g | |
| Leuco Malachite Green |
| 129-73-7 | |
| MFCD00008315 | |
| WZKXBGJNNCGHIC-UHFFFAOYSA-N | |
| 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline | |
| 67215 | |
| ≥98.0% (T) | |
| Crystal/Powder |
Safety and Handling
RTECSNumber : CY1203055
TSCA : Yes