missing translation for 'onlineSavingsMsg'
Learn More

Leuco Malachite Green 98.0+%, TCI America™

Catalog No. B04191G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04191G 1 g
B041925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B04191G Supplier TCI America Supplier No. B04191G

Chemical Identifiers

CAS 129-73-7
Molecular Formula C23H26N2
Molecular Weight (g/mol) 330.475
MDL Number MFCD00008315
InChI Key WZKXBGJNNCGHIC-UHFFFAOYSA-N
PubChem CID 67215
IUPAC Name 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C

Specifications

Melting Point 102°C
Synonym 4,4′C-Benzylidenebis(N,N-dimethylaniline), 4,4′C-Bis(dimethylamino)triphenylmethane, 4,4′C-Tetramethyldiaminotriphenylmethane
Formula Weight 330.48
Percent Purity ≥98.0% (T)
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Leuco Malachite Green
RTECSNumber CY1203055
TSCA Yes