missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leuco Crystal Violet, 99%, pure

CAS: 603-48-5 | C25H31N3 | 373.54 g/mol

$56.19 - $174.15

Chemical Identifiers

CAS 603-48-5
Molecular Formula C25H31N3
Molecular Weight (g/mol) 373.54
MDL Number MFCD00008314
InChI Key OAZWDJGLIYNYMU-UHFFFAOYSA-N
Synonym 4,4',4''-Methylidynetris(N,N-dimethylaniline), LCV
PubChem CID 69048
IUPAC Name 4-[bis[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC204330050
SDS View Documents
Thermo Scientific Chemicals
204330050
5 g Glass bottle
Each for $56.19
Only null left
 
AC204330250
SDS View Documents
Thermo Scientific Chemicals
204330250
25 g Glass bottle
Each for $174.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

603-48-5
373.54
OAZWDJGLIYNYMU-UHFFFAOYSA-N
69048
CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
C25H31N3
MFCD00008314
4,4',4''-Methylidynetris(N,N-dimethylaniline), LCV
4-[bis[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline

Specifications

175.0°C to 180.0°C
99%
CH[C6H4N(CH3)2]3
13, 315
OAZWDJGLIYNYMU-UHFFFAOYSA-N
CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
373.54
373.53
Pure
Gray to White
Powder
603-48-5
C25H31N3
MFCD00008314
4,4',4''-Methylidynetris(N,N-dimethylaniline), LCV
Authentic
4-[bis[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
69048
99%
Glass bottle
5 g
Leuco Crystal Violet, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 210-043-9

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only