missing translation for 'onlineSavingsMsg'
Learn More

Leucine, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603783
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603783 100 g Poly Bottle
18603784 250 g Poly Bottle
18603785 1 kg Poly Bottle
18603786 5 kg Poly Pail
18603787 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603783 Supplier Spectrum Chemical Mfg Cor Supplier No. L1202100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

L1202, 61-90-5, C6H13NO2

Spectrum™ Chemical Leucine, USP is a hydrophobic essential amino acid that is a component in astacin, ferritin and other buffer proteins. It is used for the creation of sterols in the muscle and adipose tissues. Widely used in sport supplements for its muscle building properties, it is also used as a food additive and flavor enhancer. Spectrum Chemical manufactured USP LEUCINE products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Chloride 0.05%
Ignition Residue 0.4%
Loss on Drying 0.2%
Packaging Poly Bottle
Specific Rotation +14.9° to +17.3°
Sulfate 0.03%
Quantity 100 g
Percent Purity 98.5 to 101.5%
Grade USP