missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ [Leu8] Renin Substrate Tetradecapeptide, Rat
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | White to off-white |
| Format | Lyophilized |
| Molecular Weight (g/mol) | 1789.06 |
| Storage Requirements | -30° to -10°C |
| Substrate Quantity | 5mg |
RUO – Research Use Only