missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Leptin Recombinant Protein

Catalog No. PIRP10936
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP10936 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP10936 Supplier Invitrogen™ Supplier No. RP10936
Only null left

Recombinant Protein

This is a recombinant protein suitable for use as a control. This protein is soluble in water and most aqueous buffers above and below the isoelectric point.

Leptin is a 167 amino acid long protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is approximately 16 kDa in mass and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, which fits with the idea that body weight is sensed as the total mass of fat in the body. Smaller amounts of leptin are also secreted by cells in the epithelium of the stomach and in the placenta. Leptin receptors are highly expressed in areas of the hypothalamus known to be important in regulating body weight, as well as in T lymphocytes and vascular endothelial cells. Leptin's effects on body weight are mediated through effects on hypothalamic centers that control feeding behavior and hunger, body temperature and energy expenditure. Leptin is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, angiogenesis, wound healing, inflammation, immune responses, influences pubertal development and fetal growth. Studies have investigated the role of leptin in obesity, anorexia nervosa, insulin resistance, and hypertension. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production.

Specifications

Accession Number P41159
For Use With (Application) Control
Formulation 45 mM sodium bicarbonate with no preservative
Gene ID (Entrez) 3952
Molecular Weight (g/mol) 16.2 kDa
Name Human Leptin
Quantity 5 mg
Source E. coli
Regulatory Status RUO
Gene Alias CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin
Common Name Leptin
Gene Symbol LEP
Biological Activity ED50 = 0.5 - 1.6 ng/mL; determined by the dose-dependent stimulation of human OB-R transfected BaF3 indicator cells.
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Human Leptin recombinant protein contains 147 amino acids
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Show More Show Less