missing translation for 'onlineSavingsMsg'
Learn More

Lepidine 97.0+%, TCI America™

Catalog No. L002425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L002425ML 25 mL
L0024100ML 100 mL
L0024500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L002425ML Supplier TCI America Supplier No. L002425ML

Chemical Identifiers

CAS 491-35-0
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00006784
InChI Key MUDSDYNRBDKLGK-UHFFFAOYSA-N
Synonym lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
PubChem CID 10285
ChEBI CHEBI:48983
IUPAC Name 4-methylquinoline
SMILES CC1=CC=NC2=CC=CC=C12

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 265°C
Quantity 25 mL
Formula Weight 143.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Lepidine
EINECSNumber (5)-0796
RTECSNumber OH0316000
TSCA Yes