Learn More
Lepidine, 97%
CAS: 491-35-0 | C10H9N | 143.189 g/mol
$83.49 - $1421.07
Chemical Identifiers
| CAS | 491-35-0 |
|---|---|
| Molecular Formula | C10H9N |
| Molecular Weight (g/mol) | 143.189 |
| MDL Number | MFCD00006784 |
| InChI Key | MUDSDYNRBDKLGK-UHFFFAOYSA-N |
| Synonym | lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894 |
| PubChem CID | 10285 |
| ChEBI | CHEBI:48983 |
| IUPAC Name | 4-methylquinoline |
| SMILES | CC1=CC=NC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1404009
|
Thermo Scientific Chemicals
A1404009 |
10 g |
Each for $83.49
|
|
|||||
|
AAA1404018
|
Thermo Scientific Chemicals
A1404018 |
50 g |
Each for $330.11
|
|
|||||
|
AAA1404030
|
Thermo Scientific Chemicals
A1404030 |
250 g |
Each for $1,421.07
|
|
|||||
Description
Lepidine is used in the preparation of certain dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLepidine is used in the preparation of certain dyes.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents. Incompatible with oxidizing agents.
Chemical Identifiers
| 491-35-0 | |
| 143.189 | |
| MUDSDYNRBDKLGK-UHFFFAOYSA-N | |
| 10285 | |
| 4-methylquinoline |
| C10H9N | |
| MFCD00006784 | |
| lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894 | |
| CHEBI:48983 | |
| CC1=CC=NC2=CC=CC=C12 |
Specifications
| 491-35-0 | |
| 1.083 | |
| >110°C (230°F) | |
| 1.62 | |
| 10 g | |
| 14,5441 | |
| Slightly soluble in water. | |
| CC1=CC=NC2=CC=CC=C12 | |
| 143.189 | |
| CHEBI:48983 | |
| 98% |
| 9°C to 10°C | |
| 260°C to 263°C | |
| C10H9N | |
| MFCD00006784 | |
| 110926 | |
| lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894 | |
| MUDSDYNRBDKLGK-UHFFFAOYSA-N | |
| 4-methylquinoline | |
| 10285 | |
| 143.19 | |
| Lepidine |
Safety and Handling
GHS H Statement
H341-H315-H319
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H341
EINECSNumber : 207-734-2
RTECSNumber : OH0316000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only