missing translation for 'onlineSavingsMsg'
Learn More

Lepidine, 97%

Catalog No. AAA1404009
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1404009 10 g
AAA1404018 50 g
AAA1404030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1404009 Supplier Thermo Scientific Chemicals Supplier No. A1404009

CAS: 491-35-0 | C10H9N | 143.189 g/mol

Lepidine is used in the preparation of certain dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lepidine is used in the preparation of certain dyes.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 491-35-0
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00006784
InChI Key MUDSDYNRBDKLGK-UHFFFAOYSA-N
Synonym lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
PubChem CID 10285
ChEBI CHEBI:48983
IUPAC Name 4-methylquinoline
SMILES CC1=CC=NC2=CC=CC=C12

Specifications

Melting Point 9°C to 10°C
Density 1.083
Boiling Point 260°C to 263°C
Flash Point >110°C (230°F)
Refractive Index 1.62
Quantity 10 g
Beilstein 110926
Merck Index 14,5441
Solubility Information Slightly soluble in water.
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material Lepidine
Show More Show Less
Hazard Category H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 207-734-2
RTECSNumber OH0316000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only