missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Lenz Law Kit

Catalog No. S12911
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S12911 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12911 Supplier Eisco™ Supplier No. LZLWKIT

Includes

Lenz law apparatus, dynamometer, two magnets, instructions

Demonstrates concepts involving Farady law, magnetism and eddy currents.

  • Pipes of differing materials demonstrate magnet braking force
  • Two magnets simultaneously dropped through aluminum and clear plastic tubes
  • Dynamometer attached to apparatus
  • Students measure additional force as magnet drops through conductive tube

Specifications

For Use With (Application) Demonstration of Farady law, magnetism and eddy currents
Grade 9 to 12
Product Type Lenz Law Kit
Includes Lenz law apparatus, dynamometer, two magnets, instructions