missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus, human HPRT, with GFP

Catalog No. A32060
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32060 100 μL
A32830 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. A32060 Supplier Invitrogen™ Supplier No. A32060

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Research Use Only. Not for use in diagnostic procedures.

Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system.

Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%.

The Invitrogen™ LentiArray CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).

Recommended Storage

Store at -68 to -85°C

Specifications

Content And Storage 1 x 100 μL. Store (at -68°C to -85°C.)
Delivery Type Lentiviral
Format Tube
Control Type Positive Control
RNAi Type gRNA
Species Human
For Use With (Application) Genome Editing
Product Line LentiArray
Product Type gRNA
Promoter U6
Quantity 100 μL
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.