missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, human, non-targeting

Catalog No. A32327
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32327 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. A32327 Supplier Invitrogen™ Supplier No. A32327

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Research Use Only. Not for use in diagnostic procedures.

Negative control gRNAs are essential in screening applications for setting thresholds for hit determination.

The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome.

Recommended Storage

Store at -68 to -85°C

Specifications

Content And Storage 1 x 1 mL. Store at -68 to -85°C.
Format Tube
Control Type Negative Control
RNAi Type gRNA
Species Human
For Use With (Application) Genome Editing
Product Line LentiArray
Product Type gRNA
Quantity 1 mL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.