missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lens Culinaris (Edible Lentil) Agglutinin (LCA), fluorescein (FITC)

Catalog No. L32475
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L32475 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L32475 Supplier Invitrogen™ Supplier No. L32475
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lens Culinaris Agglutinin (LCA), fluorescein ( FITC), is a bright-green LCA-fluorophore conjugate that is sourced from a common edible lentil.

LCA specificity is similar to concanavalin A, but narrower, resulting in differences in patterns of tissue staining. Wheat germ agglutinin, concanavalin A, and LCA are known to block inhibitory effects of nerve growth factor in cell-free systems.

• Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: mannose, glucose
• Inhibiting/eluting sugar: mixture of 200 mM α-methylmannoside/200 mM α-methylglucoside
• Blood group specificity: non-specific
• Unconjugated lectin molecular weight: 50 KDa
• Immunofluorescence working concentration: 5–20 μg/mL

LCA-FITC is provided at a 5 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+.

If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.

Specifications

Conjugate FITC
Quantity 5 mg
Source Lentil
Content And Storage When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after receipt.

Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Family Lectins

For Research Use Only. Not for use in diagnostic procedures.