missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Lens Cleaning Tissue Paper

Catalog No. S24530
Change view
Click to view available options
Quantity:
25 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24530 25 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24530 Supplier Eisco™ Supplier No. CH0629
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lens Cleaning Tissue Paper for cleaning lens, camera, spectrometer, microscope lenses, etc.

  • Available as pack of 25 leaves

Specifications

Product Type Tissue Paper
Color-coded No
Form Sachet Wipe
Scent Unscented
Sterility Non-sterile
Dimensions (L x W) 9 x 15 cm
For Use With (Application) cleaning lens, camera, spectrometer, microscope lenses
Quantity 25 Pk.