missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lemongrass Oil, Spectrum™ Chemical
Description
Spectrum™ Chemical Lemongrass Oil
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Quantity | 5 kg |
| Grade | Ungraded |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Lemongrass Oil |