missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Legionella Latex Test
Rapidly confirm predominant Legionella serogroups grown on selective media by latex agglutination.
$735.22 - $1,848.23
Specifications
| Content And Storage | 2°C to 8°C |
|---|---|
| Detectable Analytes | Legionella |
| Sample Type | Environmental Source |
| Type | Legionella Latex Test Kit |
| Quantity | 50 Tests/Kit |
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
|
OXDR0800M
|
Thermo Scientific™
DR0800M |
Legionella Latex Test |
Pack of 50 Each for $1,848.23
|
|
|||||
|
OXDR0801M
|
Thermo Scientific™
DR0801M |
Serogroup 1 Test Reagent |
Each for $735.22
|
|
|||||
|
OXDR0802M
|
Thermo Scientific™
DR0802M |
Serogroup 2-14 Test Reagent |
Pack of 50 Each for $766.80
|
|
|||||
Description
Rapidly differentiate and confirm the identity of predominant Legionella serogroups grown on selective media by latex agglutination using Thermo Scientific™ Legionella Latex Test.
Use this latex agglutination test for the identification of predominant Legionella species grown on plated media from patients with suspected Legionellosis or from environmental sources. The test allows separate identification of Legionella pneumophila serogroup 1 and serogroups 2-14 and detection of seven other Legionella species which have been implicated in human disease.Blue latex particles sensitised with antibody agglutinate in the presence of specific Legionella cell wall antigens to form visible clumps.
The following products are available for use with the Legionella Latex Test and are sold separately:
Legionella Pneumophila Serogroup 1 Test Reagent (Part No. DR0801M)
Legionella Pneumophila Serogroup 2-14 Test Reagent (Part No. DR0802M)
Legionella Species Test Reagent (Part No. DR0800M)—Legionella longbeachae 1 & 2, Legionella bozemanii 1 & 2, Legionella dumoffii, Legionella gormanii, Legionella jordanis, Legionella micdadei and Legionella anisa
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
| 2°C to 8°C | |
| Environmental Source | |
| 50 Tests/Kit |
| Legionella | |
| Legionella Latex Test Kit |