Learn More
Mopec Left Approach Wall Mount Autopsy Sink
Save time and space with a wall mounted autopsy dissection sink. Mopec™ Left Approach Wall Mount Autopsy Sink features cart trays that overhang the sink to allow for proper fluid drainage.
Supplier:  Mopec CB100
Catalog No. 22444013
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Stainless steel construction
  • Hydro aspirator with reversing flow valve, hose hanger and 8 foot clear PVC tubing
  • Cold water body rinse with vacuum breaker and serrated hose fitting
  • Hot and cold water fixture with wrist blade handles and 8 in. swing spout with vacuum breaker
  • Two interchangeable removable perforated grid plates, stainless steel construction with .5 in. recess and .75 in. diameter holes on 2 in. square centers
  • Rinse basket grid plate of stainless steel construction
  • GFCI duplex outlet with waterproof cover
  • Instrument drawer flush to the table when closed and fitted with flush inset handles at the top to keep fluid from accumulating in the drawers
  • J hook to hold hand piece
  • Concealed pressure control/shut off
  • Autopsy cart latch
  • All plumbing and electrical work accomplished at factory
  • Only trade hook up required
Warranty
1 Year Warranty
Specifications
Specifications
Autopsy Sink
71.12 cm
61 in.
213.36 cm
28 in.
84 in.
154.94 cm
Product Suggestions
Product Suggestions