missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sealed Overhead LED Light Fixture and Frame For Workstations

Catalog No. p-6899069
Change view
Click to view available options
For Use With (Equipment):
4 ft. Workstation
5 ft. Workstation
6 ft. Workstation
Wattage:
34 W
64 W
116 W
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Wattage
25100354 4 ft. Workstation 34 W
25100355 5 ft. Workstation 64 W
25100356 6 ft. Workstation 116 W
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 25100354 Supplier Fisherbrand™ Supplier No. LFA48 + SLD24
Only null left

Sealed overhead LED light and frame are ideal for 60 to 72 in. benches.

For 48, 60, or 72 in. Long Table, Instant Maximum Brightness, Cleanroom rated, Long lasting 12V LED light, creates less heat with less power consumption, Bayer polycrabonate housing for maximum difused light and long life, 5,000 Kelvin Color Temperature - similar to Fluorescent Daylight, Quiet and no-fluorescent-buzzing sound, Sealed to eliminate dust and reduce fire hazards, Stainless steel mounting brackets, No high voltage danger, 6A 115V power supply, 10 year manufacturer warranty

  • For 48 in., 60 in., or 72 in. Long Tables
  • Instant Maximum Brightness
  • Cleanroom rated
  • Long lasting 12V LED light, creates less heat with less power consumption
  • Bayer polycarbonate housing for maximum difused light and long life
  • 5,000 Kelvin Color Temperature - similar to Fluorescent Daylight
  • Quiet and no-fluorescent-buzzing sound
  • Sealed to eliminate dust and reduce fire hazards
  • Stainless steel mounting brackets
  • No high voltage danger
  • 6A 115V power supply
  • 10 year manufacturer warranty

Order Info

Must order upright post (Fisher SKU 25100350)

Warranty

25-year manufacturer warranty

Specifications

Light Source LED
Voltage 12 V
Wattage 34 W
Amperage 6 A
Case Material Polycarbonate, Stainless Steel
Description 48 in. light frame; 24 in. light fixture
For Use With (Application) Workstation lighting
For Use With (Equipment) 4 ft. Workstation
Includes Switch and 6 A, 115 V power supply cord
Product Type Light Fixture and Frame