missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek LED Cubes for Imaging Multi-Mode Reader
GREENER_CHOICE

Catalog No. BT1225001
Change view
Click to view available options
Light Source:
390 nm LED cube
655 nm LED cube
CY5 LED
CY7 LED
GFP/CFP LED
RFP LED
Texas Red LED
YFP LED
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Light Source
BT1225001 GFP/CFP LED
BT1225004 YFP LED
BT1225005 CY5 LED
BT1225003 RFP LED
BT1225002 Texas Red LED
BT1225006 CY7 LED
BT1225008 655 nm LED cube
BT1225009 390 nm LED cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. BT1225001 Supplier Agilent BioTek Supplier No. 1225001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Imaging Multi-Mode Readers.

For use with Imaging Multi-Mode Readers.

Specifications

Type LED Light Cube
For Use With (Equipment) Imaging Multi-Mode Reader
Color Green/Cyan
Light Source GFP/CFP LED