missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCI America™ Lectin, Fucose specific from Aspergillus oryzae (5mg/mL, PBS pH6.5)
Specifications
Specifications
| Concentration | 4.5 to 5.5 mg/mL |
| Molecular Weight (g/mol) | 30000∽40000 (SDS-PAGE) |
| Storage Requirements | 0-10°C |
| Form | Liquid |