missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lecithin, Refined

Catalog No. p-4408341
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3648630 250 g
AA36486A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3648630 Supplier Thermo Scientific Chemicals Supplier No. 03648630
Only null left

CAS: 8002-43-5 | C42H80NO8P | 758.075 g/mol

Lecithin is used as an emulsifying and stabilizing agent in the food, pharmaceutical, and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacturing of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
MDL Number MFCD00147406
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Quantity 250 g
Beilstein 5209585
Solubility Information Soluble in chloroform. Very partially soluble in water.
Physical Form Powder
Chemical Name or Material Lecithin
Hazard Category H373
Precautionary Statement P260-P314-P501c
EINECSNumber 232-307-2
RTECSNumber OG7565000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only