missing translation for 'onlineSavingsMsg'
Learn More

Lecithin, Granular, Food Grade, Spectrum™ Chemical

Catalog No. p-7157681
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603767 500 g Poly Bottle
18603768 2.5 kg Poly Pail
18603769 12 kg Fiber Drum
18603770 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603767 Supplier Spectrum Chemical Mfg Cor Supplier No. L1617500GM
Only null left

L1617, 8002-43-5

Spectrum™ Chemical Lecithin, Granular, Food Grade is used for a host of applications such as food additive, dietary supplement, stabilizing agent and dispersing agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.08
InChI Key JLPULHDHAOZNQI-UHFFFAOYNA-N
IUPAC Name (2-{[3-(hexadecanoyloxy)-2-(octadeca-9,12-dienoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Grade Food Grade