missing translation for 'onlineSavingsMsg'
Learn More

Measuring Spoons

Catalog No. S95513
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95513 K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95513 Supplier Customer Channels Group (Ccg) Supplier No. LER4291

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

0.12 teaspoon (0.62mL), 0.25 teaspoon (1.25mL), 0.5 teaspoon (2.5mL), 1 teaspoon (5mL), 0.5 tablespoon (7.5mL), and 1 tablespoon (15mL) sizes

For age group between 5-13 years.

  • Set of six
  • Sturdy plastic
  • With detachable rings and nests for storage

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Format Hands-on Learning
Product Type Measuring Spoon Set
Includes 0.12 Teaspoon (0.62mL), 0.25 teaspoon (1.25mL), 0.5 teaspoon (2.5mL), 1 teaspoon (5mL), 0.5 tablespoon (7.5mL) and 1 tablespoon (15mL) sizes
For Use With (Application) Easy storage
Subject Mathematics