missing translation for 'onlineSavingsMsg'
Learn More

Measuring Cups

Catalog No. S8820A
Change view
Click to view available options
Grade:
K to 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S8820A K to 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S8820A Supplier Customer Channels Group (Ccg) Supplier No. LER4290

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

0.12 cup (29.5mL), 0.25 cup (59mL), 0.3 cup (79mL), 0.5 cup (118mL), and 1 cup (236mL) sizes

For age group between 5-13 years.

  • Set of five
  • Sturdy plastic
  • With detachable rings and nests for storage

Specifications

Grade Level Elementary
Grade K to 2
Format Hands-on Learning
Product Type Measuring Cup Set
Includes 0.12 Cup (29.5mL), 0.25 cup (59mL), 0.3 cup (79mL), 0.5 cup (118mL) and 1 cup (236mL) sizes
For Use With (Application) Storage
Subject Mathematics