missing translation for 'onlineSavingsMsg'
Learn More

Lead Tin, solder alloy, 1.6mm (0.06in) dia, with rosin core

Catalog No. AA41024TB
Change view
Click to view available options
Quantity:
113 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41024TB 113 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41024TB Supplier Thermo Scientific Chemicals Supplier No. 041024TB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Lead Tin, Solder Alloy
Name Note 1.6mm (0.06 in.) dia., with rosin core
Avg. Mol. Wt. or Mol. Wt. Range Pb:Sn; 50:50 wt%
Quantity 113 g
Form Solid
MDL Number MFCD00798549

RUO – Research Use Only