missing translation for 'onlineSavingsMsg'
Learn More

Lead Tetraacetate (contains Acetic Acid) 96.0+%, TCI America™

Catalog No. p-7131778
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L002125G 25 g
L0021500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L002125G Supplier TCI America Supplier No. L002125G
Only null left

Chemical Identifiers

CAS 546-67-8
Molecular Formula C8H16O8Pb
Molecular Weight (g/mol) 447.408
MDL Number MFCD00008693
InChI Key NVTAREBLATURGT-UHFFFAOYSA-N
Synonym lead iv acetate
PubChem CID 50931538
IUPAC Name acetic acid;lead
SMILES CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pb]

Specifications

Melting Point 180°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3087
Formula Weight 443.38
Percent Purity ≥96.0% (T)
Chemical Name or Material Lead Tetraacetate (contains Acetic Acid)
EINECSNumber (2)-0693
RTECSNumber AI5300000
TSCA Yes
Recommended Storage Refrigerator