Learn More
LaMotteā„¢ Lead Screening Test Kit
An affordable, safe, and reliable screening test to identify lead in drinking water. Ideal for classroom studies or the professional screenng multiple tests on campuses, office buildings, etc.
Supplier:  LaMotteā„¢ EMD50140
Catalog No. S29054
Only null left
Explore available promotions
Includes: 10 test packs and diagrammed instructions
Description
Description
In 10 minutes the analyst can detect lead levels above 15 ppb—the EPA action level. Just immerse a strip in a water sample and read after a 10 minute waiting period.
  • This multi-test package is designed for the professional monitoring multiple sites
  • The kit includes 10 test packs with one diagrammed instruction
  • Each test comes in a sealed pouch with a test vial and dropper
  • Simple test protocol: immerse, wait 10 minutes, and read Easy analysis—1 Line or 2 Line result indicates result Detects Lead to EPA action level of 15 parts per billion (ppb)
Specifications
Specifications
Water test Kit
10 test packs and instructions
Product Suggestions
Product Suggestions