missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Subacetate, Spectrum™ Chemical
$2534.79 - $14036.50
Chemical Identifiers
| CAS | 1335-32-6 |
|---|---|
| Molecular Formula | C4H10O8Pb3 |
| Molecular Weight (g/mol) | 807.70 |
| InChI Key | VLOJXAQYHIVPFI-UHFFFAOYSA-H |
| IUPAC Name | tris(λ²-lead(2+)) diacetate tetrahydroxide |
| SMILES | [OH-].[OH-].[OH-].[OH-].[Pb++].[Pb++].[Pb++].CC([O-])=O.CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18611256
|
Spectrum Chemical Mfg Cor
L108825KG |
2.5 kg | Amber Glass Bottle |
Each for $2,534.79
|
|
||||
|
18611255
|
Spectrum Chemical Mfg Cor
L108812KG |
12 kg | Poly Pail |
Each for $8,564.82
|
|
||||
|
18611257
|
Spectrum Chemical Mfg Cor
L108850KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 1335-32-6 | |
| 807.70 | |
| tris(λ²-lead(2+)) diacetate tetrahydroxide |
| C4H10O8Pb3 | |
| VLOJXAQYHIVPFI-UHFFFAOYSA-H | |
| [OH-].[OH-].[OH-].[OH-].[Pb++].[Pb++].[Pb++].CC([O-])=O.CC([O-])=O |
Specifications
| 1335-32-6 | |
| 2.5 kg | |
| VLOJXAQYHIVPFI-UHFFFAOYSA-H | |
| tris(λ²-lead(2+)) diacetate tetrahydroxide | |
| Reagent |
| 1 | |
| C4H10O8Pb3 | |
| [OH-].[OH-].[OH-].[OH-].[Pb++].[Pb++].[Pb++].CC([O-])=O.CC([O-])=O | |
| 807.70 | |
| Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.