missing translation for 'onlineSavingsMsg'
Learn More

Lead rod, 12.7mm (0.5in) dia, Puratronic™, 99.9998% (metals basis)

Catalog No. AA1134618
Change view
Click to view available options
Quantity:
50 g
210 g
4 x 210 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1134618 50 g
AA1134697 210 g
AA1134698 4 x 210 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1134618 Supplier Thermo Scientific Chemicals Supplier No. 01134618

CAS: 7439-92-1 | Pb | 207.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Quantity 50 g
Odor Odorless
Weight ≈3.5cm/50g
Form Rod
Percent Purity 99.9998%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Lead rod, 12.7mm (0.5 in.) dia., May be in multiple pieces
Hazard Category H350-H360FD-H362-H372
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
EINECSNumber 231-100-4
RTECSNumber OF7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only