Learn More
Lead powder, -200 mesh, 99% (metals basis)
CAS: 7439-92-1 | Pb | 207.20 g/mol
Supplier: Thermo Scientific Chemicals 00094236
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Black to Gray | |
| 7439-92-1 | |
| 500 g | |
| Odorless | |
| Powder | |
| MFCD00134050 | |
| WABPQHHGFIMREM-UHFFFAOYSA-N | |
| lead | |
| 5352425 |
| Lead | |
| Gray to black powder | |
| -200 Mesh | |
| 99% (Metals basis) | |
| Pb | |
| plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
| [Pb] | |
| 207.20 | |
| CHEBI:27889 |
Chemical Identifiers
| 7439-92-1 | |
| 207.20 | |
| WABPQHHGFIMREM-UHFFFAOYSA-N | |
| 5352425 | |
| lead |
| Pb | |
| MFCD00134050 | |
| plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
| CHEBI:27889 | |
| [Pb] |
Safety and Handling
P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
H350-H360FD-H362-H372
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 231-100-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only