missing translation for 'onlineSavingsMsg'
Learn More

Lead Oxide, Yellow Powder, 95%, Spectrum™ Chemical

Catalog No. p-7165777
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18611249 125 g Poly Bottle
18611251 500 g Poly Bottle
18611248 2.5 kg Poly Bottle
18611250 12 kg Poly Pail
18611247 45 kg Fiber Drum
5 options

Catalog No. 18611249

Supplier: Spectrum Chemical Mfg Cor L1080125GM

Only null left

Chemical Identifiers

CAS 1317-36-8

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 95%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.