missing translation for 'onlineSavingsMsg'
Learn More

Lead Oxide, Yellow Powder, 95%, Spectrum™ Chemical

Catalog No. 18611249
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611249 125 g Poly Bottle
18611251 500 g Poly Bottle
18611248 2.5 kg Poly Bottle
18611250 12 kg Poly Pail
18611247 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611249 Supplier Spectrum Chemical Mfg Cor Supplier No. L1080125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 1317-36-8

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 95%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.