missing translation for 'onlineSavingsMsg'
Learn More

Lead Oxide, Red Powder, 98%, Spectrum™ Chemical

Catalog No. 18611246
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611246 45 kg Fiber Drum
18611242 125 g Amber Glass Bottle
18611244 500 g Amber Glass Bottle
18611245 2.5 kg Amber Glass Bottle
18611243 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
Catalog No. 18611246 missing translation for 'mfr' Spectrum Chemical Mfg Cor missing translation for 'supplierNo' L108545KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1314-41-6
Molecular Formula O4Pb3
Molecular Weight (g/mol) 685.60
InChI Key XMFOQHDPRMAJNU-UHFFFAOYSA-N
IUPAC Name bicyclo[3.1.1]triplumboxan-1-yl
SMILES O1[Pb]2O[Pb]1O[Pb]O2

Specifications

CAS Max % 1
Quantity 45 kg
Percent Purity 98%
Grade Reagent
Packaging Fiber Drum

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.