missing translation for 'onlineSavingsMsg'
Learn More

Lead Oxide, Red Powder, 98%, Spectrum™ Chemical

Catalog No. 18611242
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611242 125 g Amber Glass Bottle
18611244 500 g Amber Glass Bottle
18611245 2.5 kg Amber Glass Bottle
18611243 12 kg Poly Pail
18611246 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611242 Supplier Spectrum Chemical Mfg Cor Supplier No. L1085125GM

Chemical Identifiers

CAS 1314-41-6
Molecular Formula O4Pb3
Molecular Weight (g/mol) 685.60
InChI Key XMFOQHDPRMAJNU-UHFFFAOYSA-N
IUPAC Name bicyclo[3.1.1]triplumboxan-1-yl
SMILES O1[Pb]2O[Pb]1O[Pb]O2

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.