Learn More
Lead(IV) oxide, 97%
CAS: 1309-60-0 | O2Pb | 239.20 g/mol
$218.04 - $822.51
Chemical Identifiers
| CAS | 1309-60-0 |
|---|---|
| Molecular Formula | O2Pb |
| Molecular Weight (g/mol) | 239.20 |
| MDL Number | MFCD00011165 |
| InChI Key | YADSGOSSYOOKMP-UHFFFAOYSA-N |
| Synonym | lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french |
| PubChem CID | 14793 |
| IUPAC Name | dioxolead |
| SMILES | O=[Pb]=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1274236
|
Thermo Scientific Chemicals
A1274236 |
500 g |
Each for $218.04
|
|
|||||
|
AAA127420E
|
Thermo Scientific Chemicals
A127420E |
2500 g |
Each for $822.51
|
|
|||||
Description
Lead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.
Solubility
Insoluble in water. Soluble in dil.HCl.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 1309-60-0 | |
| 239.20 | |
| YADSGOSSYOOKMP-UHFFFAOYSA-N | |
| 14793 | |
| O=[Pb]=O |
| O2Pb | |
| MFCD00011165 | |
| lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french | |
| dioxolead |
Specifications
| 290°C | |
| 500 g | |
| MFCD00011165 | |
| 14,5407 | |
| Insoluble in water. Soluble in dil.HCl. | |
| O=[Pb]=O | |
| 239.20 | |
| 239.19 | |
| Odorless | |
| Lead(IV) oxide |
| 1309-60-0 | |
| O2Pb | |
| UN1872 | |
| lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french | |
| YADSGOSSYOOKMP-UHFFFAOYSA-N | |
| dioxolead | |
| 14793 | |
| 97% | |
| 9.38 |
Safety and Handling
GHS H Statement
H360-H373-H272-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c
H272-H302+H332-H350-H360Df-H373
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: LEAD DIOXIDE
EINECSNumber : 215-174-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only