missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) oxide, 97%

Catalog No. AAA1274236
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1274236 500 g
AAA127420E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1274236 Supplier Thermo Scientific Chemicals Supplier No. A1274236
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 1309-60-0 | O2Pb | 239.20 g/mol

Lead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.

Solubility
Insoluble in water. Soluble in dil.HCl.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
IUPAC Name dioxolead
SMILES O=[Pb]=O

Specifications

Melting Point 290°C
Quantity 500 g
UN Number UN1872
Merck Index 14,5407
Solubility Information Insoluble in water. Soluble in dil.HCl.
Formula Weight 239.19
Percent Purity 97%
Odor Odorless
Density 9.38
Chemical Name or Material Lead(IV) oxide
Hazard Category H272-H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H272-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: LEAD DIOXIDE
EINECSNumber 215-174-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only