missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) oxide, 97+%, extra pure

Catalog No. AC217530050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217530050 5 g Glass bottle
AC217531000 100 g Glass bottle
AC217535000 500 g Glass bottle
AC217530025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC217530050 Supplier Thermo Scientific Chemicals Supplier No. 217530050

CAS: 1309-60-0 | O2Pb | 239.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
SMILES O=[Pb]=O

Specifications

Melting Point 290°C
Color Brown to Black
Physical Form Powder
Quantity 5 g
Assay Percent Range 97+%
Linear Formula PbO2
Fieser 01,5331; 02,233; 03,168; 11,282
Merck Index 15, 5462
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hcl and glacial acetic acid,insoluble in alcohol
IUPAC Name dioxolead
Formula Weight 239.19
Percent Purity ≥97%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Lead(IV) oxide, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child.
Suspected of damaging fertility.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C
EINECSNumber 215-174-5
RTECSNumber OG0700000
TSCA TSCA

RUO – Research Use Only