missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) fluoride, 99%

Catalog No. AA1307004
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1307004 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1307004 Supplier Thermo Scientific Chemicals Supplier No. 01307004
Only null left

CAS: 7783-59-7 | F4Pb | 283.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-59-7
Molecular Formula F4Pb
Molecular Weight (g/mol) 283.194
MDL Number MFCD00016278
InChI Key YAFKGUAJYKXPDI-UHFFFAOYSA-J
Synonym lead iv fluoride, lead tetrafluoride, pbf4, tetrafluoridolead, blei iv-fluorid
PubChem CID 123258
ChEBI CHEBI:37186
IUPAC Name tetrafluoroplumbane
SMILES F[Pb](F)(F)F

Specifications

Physical Form Crystalline
Quantity 2 g
UN Number UN2291
Formula Weight 283.18
Percent Purity 99%
Odor Odorless
Sensitivity Moisture sensitive
Chemical Name or Material Lead(IV) fluoride
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber 232-012-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only