missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead ingot, 99.999% (metals basis)
CAS: 7439-92-1 | Pb | 207.20 g/mol
$575.49 - $575.49
Chemical Identifiers
| CAS | 7439-92-1 |
|---|---|
| Molecular Formula | Pb |
| Molecular Weight (g/mol) | 207.20 |
| MDL Number | MFCD00134050 |
| InChI Key | WABPQHHGFIMREM-UHFFFAOYSA-N |
| Synonym | plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant |
| PubChem CID | 5352425 |
| ChEBI | CHEBI:27889 |
| IUPAC Name | lead |
| SMILES | [Pb] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7439-92-1 | |
| 207.20 | |
| WABPQHHGFIMREM-UHFFFAOYSA-N | |
| 5352425 | |
| lead |
| Pb | |
| MFCD00134050 | |
| plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
| CHEBI:27889 | |
| [Pb] |
Specifications
| Gray | |
| Gray ingot | |
| Odorless | |
| Ingot | |
| MFCD00134050 | |
| WABPQHHGFIMREM-UHFFFAOYSA-N | |
| lead | |
| 5352425 | |
| Lead |
| 7439-92-1 | |
| 100 g | |
| 99.999% (Metals basis) | |
| Pb | |
| plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
| [Pb] | |
| 207.20 | |
| CHEBI:27889 |
Safety and Handling
P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
H350-H360FD-H362-H372
EINECSNumber : 231-100-4
RTECSNumber : OF7525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only