missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) zirconium oxide, 99.7% (metals basis excluding Hf), Hf 1% max, Thermo Scientific Chemicals
Description
It is practically applied in the area of electroceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 100 g |
| UN Number | UN3077 |
| Solubility Information | Insoluble in all solvents. |
| Formula Weight | 346.42 |
| Percent Purity | 99.7% |
| Odor | Odorless |
| Density | 7.0 g/mL |
| Assay | (metals basis excluding Hf), Hf 1% max. |
| Chemical Name or Material | Lead(II) zirconium oxide |
Safety and Handling
| Hazard Category | H302+H332-H350-H360Df-H373 |
| Hazard Statement | GHS H Statement H360-H373-H302-H332 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 235-039-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only