missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) titanium oxide, 99.9% (metals basis)

Catalog No. AA3567118
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3567118 50 g
AA3567130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3567118 Supplier Thermo Scientific Chemicals Supplier No. 03567118
Only null left

CAS: 12060-00-3 | O3PbTi | 303.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12060-00-3
Molecular Formula O3PbTi
Molecular Weight (g/mol) 303.064
MDL Number MFCD00011159
InChI Key NKZSPGSOXYXWQA-UHFFFAOYSA-N
Synonym lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder,
PubChem CID 16211560
IUPAC Name dioxido(oxo)titanium;lead(2+)
SMILES [O-][Ti](=O)[O-].[Pb+2]

Specifications

Color Yellow
Physical Form Powder
Quantity 50 g
UN Number UN3288
Formula Weight 303.09
Percent Purity 99.9%
Odor Odorless
Density 7.520 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) titanium oxide
Hazard Category H300-H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H300-H360-H373-H332
Fatal if swallowed.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 235-038-9
RTECSNumber XR1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only