missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) tetrafluoroborate, 50% w/w aq. soln.

Catalog No. AA41956A1
Change view
Click to view available options
Quantity:
1 kg
3 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41956A1 1 kg
AA41956D5 3 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41956A1 Supplier Thermo Scientific Chemicals Supplier No. 041956A1

Used in Tin-Lead Electroplating | CAS: 13814-96-5 | B2F8Pb | 380.80 g/mol

Used in tin-lead electroplatingLead(II) tetrafluoroborate is used as a catalyst involved in calixarene formation through hydrogen-bonded dimmers and in the preparation of linear polyesters. It is involved in the development and optimization of a bipolar lead-boron tetrafluoride accumulator. It is an electroplating solution for coating metal objects with lead and curing agent for epoxy resins. Further, it is used in printing lines, tin-lead alloy plating, lead soldering and also as a circuit board tin-lead alloy plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13814-96-5
Molecular Formula B2F8Pb
Molecular Weight (g/mol) 380.80
MDL Number MFCD00016270
InChI Key UQGXAENNKXXYNF-UHFFFAOYSA-N
Synonym lead ii tetrafluoroborate, lead fluoroborate, lead ii tetrafluoroborate solution, lead boron fluoride, lead tetrafluoroborate, unii-wax1e0vj84, lead ii fluoroborate, acmc-1bpsn, wax1e0vj84, lead 2+ ditetrafluoroborate
PubChem CID 12598180
SMILES [Pb++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 kg
Solubility Information Miscible with water.
IUPAC Name lead(2+);ditetrafluoroborate
Formula Weight 380.81
Odor Odorless
Density 1.62
Chemical Name or Material Lead(II) tetrafluoroborate
Hazard Category H302+H332-H350-H360Df-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 237-486-0
RTECSNumber ED2700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only