missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) telluride, 99.999% (metals basis)

Catalog No. p-7080449
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8828514 25 g
AA8828522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8828514 Supplier Thermo Scientific Chemicals Supplier No. 08828514
Only null left

CAS: 1314-91-6 | PbTe | 334.80 g/mol

Lead(II) telluride is used to make thermoelectric converter (cooler, generator, heat), infrared detectors and tunnel diode. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-91-6
Molecular Formula PbTe
Molecular Weight (g/mol) 334.80
MDL Number MFCD00016274
InChI Key OCGWQDWYSQAFTO-UHFFFAOYSA-N
Synonym lead telluride, lead ii telluride, unii-v1og6oa4bj, v1og6oa4bj, plumbanetellone, bleitellurid, lead monotelluride, lead ii telluride trace metals basis
PubChem CID 4389803
IUPAC Name tellanylidenelead
SMILES [Te]=[Pb]

Specifications

Melting Point 917°C
Physical Form Pieces
Quantity 25 g
UN Number UN3284
Merck Index 14,5422
Solubility Information Insoluble in water.
Formula Weight 334.79
Percent Purity 99.999%
Odor Product specific
Density 8.164 g/mL
Assay (metals basis)
Chemical Name or Material Lead (II) telluride
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-247-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only