missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) telluride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. AA3631718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3631718 50 g
AA3631709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3631718 Supplier Thermo Scientific Chemicals Supplier No. 03631718

For Use in Semiconductor Research, Photoconductor Cells

In semiconductor research, photoconductor cellsLead(II) telluride is used as an infrared detector material. It is also used in spacecraft power applications. Further, it is used in infrared detection, imaging and in solar cells. In addition, it acts as a semiconductor and photoconductor in the form of single crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Resistant to acids

Chemical Identifiers

CAS 1314-91-6
Molecular Formula PbTe
Molecular Weight (g/mol) 334.80
MDL Number MFCD00016274
InChI Key OCGWQDWYSQAFTO-UHFFFAOYSA-N
Synonym lead telluride, lead ii telluride, unii-v1og6oa4bj, v1og6oa4bj, plumbanetellone, bleitellurid, lead monotelluride, lead ii telluride trace metals basis
PubChem CID 4389803
IUPAC Name tellanylidenelead
SMILES [Te]=[Pb]

Specifications

Melting Point 917°C
Color Gray
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 334.79
Density 8.164
Chemical Name or Material Lead(II) telluride
Hazard Category H302+H332-H350-H360Df-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 215-247-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only