missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) sulfate, Puratronic™, 99.999% (metals basis)

Catalog No. p-7048516
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1073009 10 g
AA1073018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1073009 Supplier Thermo Scientific Chemicals Supplier No. 01073009
Only null left

CAS: 7446-14-2 | O4PbS | 303.256 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7446-14-2
Molecular Formula O4PbS
Molecular Weight (g/mol) 303.256
MDL Number MFCD00011166
InChI Key KEQXNNJHMWSZHK-UHFFFAOYSA-L
Synonym lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite
PubChem CID 24008
IUPAC Name lead(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Pb+2]

Specifications

Melting Point 1170°C
Physical Form Crystalline
Quantity 10 g
UN Number UN2291
Merck Index 14,5420
Formula Weight 303.25
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Density 6.2 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) sulfate
Show More Show Less
Hazard Category H302+H312+H332-H350-H360-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-198-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only