missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) selenide, 99.999+% (metals basis)

CAS: 12069-00-0 | PbSe | 286.20 g/mol

$455.45 - $1685.86

Chemical Identifiers

CAS 12069-00-0
Molecular Formula PbSe
Molecular Weight (g/mol) 286.20
MDL Number MFCD00016273
InChI Key GGYFMLJDMAMTAB-UHFFFAOYSA-N
Synonym lead selenide, lead ii selenide, lead ii selenide trace metals basis 25g
PubChem CID 61550
IUPAC Name selanylidenelead
SMILES [Se]=[Pb]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4491614
View Documents
Thermo Scientific Chemicals
04491614
25 g
Each for $455.45
Only null left
 
AA4491622
View Documents
Thermo Scientific Chemicals
04491622
100 g
Each for $1,685.86
Only null left
 
Description

Description

Lead(II) selenide is used in the production of infrared detectors for thermal imaging. It is also used as quantum dots in nanocrystal solar cell. It is an important material and finds applications in photographic plates, and photo resistors and optoelectronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12069-00-0
286.20
GGYFMLJDMAMTAB-UHFFFAOYSA-N
61550
[Se]=[Pb]
PbSe
MFCD00016273
lead selenide, lead ii selenide, lead ii selenide trace metals basis 25g
selanylidenelead

Specifications

Gray
Fused Lump
>99.999% (Metals basis)
MFCD00016273
Insoluble in water.
[Se]=[Pb]
286.20
286.15
12069-00-0
25 g
PbSe
lead selenide, lead ii selenide, lead ii selenide trace metals basis 25g
GGYFMLJDMAMTAB-UHFFFAOYSA-N
selanylidenelead
61550
Lead(II) selenide
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350-H360Df-H373

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 235-109-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only